missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ 850 Homogenizer

The Fisherbrand™ 850 Homogenizer is specifically designed to disrupt, emulsify or homogenize a wide range of samples in volumes ranging from 0.2mL to 5L. 

Brand:  Fisherbrand™

Additional Details : Weight : 7.34400kg

Product Code. 15505819

  • 1805.00 € / Each
Estimated Shipment: 13-06-2024
to see stock.
Add to basket

Description

Description

  • Powerful 850w motor
  • Integrated digital speed control, displays speed and run time
  • Variable speed control from 5,000 to 28,000rpm
  • Compatible with stainless steel and disposable generator probes
  • Processing range from 0.2mL to 5L
  • Includes motor clamp
  • Stand (baseplate and post) is sold separately; see Cat. No. 15535819

Compliance

CE Approved

Services

2 year warranty

Specifications

Specifications

Homogenizer
220 V, 850 W, EU/UK Cord
6.9 cm
Tissue homogenization, tissue disruption, protein extraction, RNA extraction, DNA extraction, metabolite extraction, emulsification, proteomics, PCR, enzyme extraction, high speed stirring, powder suspension, genomics, metabolomics
1 Ea.
1.6 kg
5000 to 28,000 rpm
0.2 mL to 5 L
21.5 cm
Homogenizer, Motor Clamp. (Stand is sold separately, 15535819.)
EU/UK Plug
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers