missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva (formerly Pall Lab) AcroPak™ 500 Capsules with Supor™ Membrane, Sterile
AcroPak capsules designed for research applications only.
166.00 € - 215.00 €
Specifications
Material (Membrane) | Hydrophilic PES |
---|---|
Sterility | Sterile |
Filtration Area | 500 cm2 |
No. per Pack | 1/Ea. |
Outlet Style | Stepped Hose Barb |
Product Code | Brand | Pore Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Pore Size | Price | Quantity & Availability | |||||
17124251
|
Cytiva (formerly Pall Lab)
12993 |
0.8/0.45 μm |
166.00 €
Each |
Estimated Shipment: 21-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17104251
|
Cytiva (formerly Pall Lab)
12991 |
0.8/0.2 μm |
166.00 €
Each |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17144251
|
Cytiva (formerly Pall Lab)
12995 |
0.2/0.2 μm |
215.00 €
Each |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17164251
|
Cytiva (formerly Pall Lab)
12997 |
0.1/0.1 μm |
215.00 €
Each |
Estimated Shipment: 03-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- For buffers, tissue culture media, and other biological fluids
- Newer technology recommended for simplified scale-up
- High filtration area and compact size are ideal for upscale trials
Specifications
Hydrophilic PES | |
500 cm2 | |
Stepped Hose Barb |
Sterile | |
1/Ea. |