missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium hydroxide solution, TraceSELECT Ultra, for ultratrace analysis,≥20% in H2O, Fluka, Honeywell
Click to view available options
Quantity:
250 mL
Description
TraceSELECT™ Ultra and TraceSELECT
- ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
- sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
- the acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (μg/kg) trace range
- packaged in high-quality containers appropriate for each product
- the reagents are produced and bottled under clean-room conditions
Specifications
Boiling Point | 24°C |
CAS Min % | ≥70.00, ≥20.00 |
CAS Max % | <90.00, <30.00 |
Beilstein | 3587154 |
Density | 0.90 g/cm3 |
Quantity | 250 mL |
Grade | Trace Analysis |
Packaging | Poly Bottle |
Physical Form | Liquid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'