Learn More
Axygen™ Black Hanging Format Automation Pipet Tips for Hamilton Microlab™ Star Line and Nimbus™
Compatible with Hamilton MICROLAB™ STAR and Nimbus workstations
Brand:  Axygen™ HTF-1000-CBK-HTR-S
Code : A0
Additional Details : Weight : 6.80388kg
 View more versions of this product
Product Code. 12832293
  • 228.00 € / Pack of 1536
Estimated Shipment: 06-06-2024
to see stock.
Add to basket
Description
Description
Axygen offers a comprehensive line of tips for Hamilton™ Microlab™ automation systems.
The tips are:
  • Manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality
  • RNase-/DNase-free and non-pyrogenic
  • Available in non-sterile, sterile, non-filtered and filtered
  • Conductive black polypropylene tips are available in 96-well hanging tip rack and nested tip rack and individual deck-seating rack format
  • Designed for Hamilton™ MICROLAB™ STAR, STARlet, STARplus and Nimbus™ workstations.
Specifications
Specifications
1000 μL
Sterile
Not Autoclavable
The conductive, liquid level sensing tips are available in four sizes and specially designed for use with Hamilton™ liquid handling platforms, they enable precision pipetting across a wide range of applications
Automation, Round
Hamilton MICROLAB STAR, Hamilton MICROLAB STARlet, Hamilton MICROLAB STARplus, Hamilton MICROLAB Nimbus
Filtered
Black
DNase, RNase and Pyrogen free
Hanging Tip Rack
1 Rack/Pk, 16 Pks/Cs.
Polypropylene