missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Azlon™ Stand for Round Bottomed Flask
Suitable for most round bottomed flasks
Brand: Azlon TWR038
26.96 EUR valid until 2024-12-31
Use promo code "23259" to get your promotional price.
Additional Details : Weight : 0.14400kg
Description
- Suitable for most round bottom flasks
- Has stepped concentric rings to help stabilise flask
- Autoclavable up to 121°C
- Has good chemical resistance
Specifications
Polypropylene | |
Flask stand | |
Translucent | |
Suitable for most round bottomed flasks |