Learn More
Texwipe™ BetaMop™ II
Low contamination properties make this the perfect mop for floor cleaning in critical environments
168.00 € - 429.00 €
Product Code | Brand | Product Type | For Use With (Equipment) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | For Use With (Equipment) | Price | Quantity & Availability | ||||
11581214
|
Texwipe™
TX7106 |
BetaMop II Mop | Wring-bucket Systems |
168.00 €
Each |
Estimated Shipment: 22-07-2024
Log in to see stock. |
||||
11541224
|
Texwipe™
TX716R |
Refill Mop Head | BetaMop II |
429.00 €
Pack of 6 |
Estimated Shipment: 24-07-2024
Log in to see stock. |
||||
Description
Durable, absorbent, corrosion-resistant, and low-contaminating; Autoclavable.
Durability, corrosion resistance, and low contamination properties make this the perfect mop for floor cleaning in critical environments. High-capacity, seamless, tubular-knit polyester mop head provides excellent absorbency. BetaMop II may be used with all wring-bucket systems. Both the mop head and clamp are autoclavable for use in sterile environments.
Specifications
Autoclavable | |
Corrosion | |
Polyester |