missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Cassette II Biopsy Tissue Cassettes in Tube Packs
Biopsy Cassettes are ideal for small tissue samples. They are compatible with all Epredia PrintMate™ Cassette Printers, have a front edge lid hinge, and loose-packed lids that are available in 12 color varieties.
131.00 € - 236.00 €
Specifications
Closure Included | Yes |
---|---|
Quantity | 750/Cs. |
Pore Style | Small Squares |
Closure Type | Separate Lid, Front Hinge |
PrintMate Compatible | Yes |
Product Code | Brand | Color | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Price | Quantity & Availability | |||||
16244322
|
Epredia™
B851730BL |
Blue |
236.00 €
Pack of 750 |
Estimated Shipment: 12-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16344066
|
Epredia™
B851730GN |
Green |
137.00 €
Pack of 750 |
Estimated Shipment: 26-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16354066
|
Epredia™
B851730GY |
Gray |
137.00 €
Pack of 750 |
Estimated Shipment: 26-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16364066
|
Epredia™
B851730LA |
Lavender |
137.00 €
Pack of 750 |
Estimated Shipment: 26-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16374066
|
Epredia™
B851730LG |
Light Green |
137.00 €
Pack of 750 |
Estimated Shipment: 26-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16384066
|
Epredia™
B851730LI |
Lilac |
137.00 €
Pack of 750 |
Estimated Shipment: 26-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16394066
|
Epredia™
B851730OR |
Orange |
137.00 €
Pack of 750 |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16304076
|
Epredia™
B851730PK |
Pink |
137.00 €
Pack of 750 |
Estimated Shipment: 26-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16324076
|
Epredia™
B851730WH |
White |
137.00 €
Pack of 750 |
Estimated Shipment: 26-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16334076
|
Epredia™
B851730YW |
Yellow |
137.00 €
Pack of 750 |
Estimated Shipment: 26-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16314076
|
Epredia™
B851730TN |
Tan |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Printing
- Surface finish and reinforced print faces are engineered to optimize print quality
- Tube design is precisely engineered to reduce jamming and prevent downtime
Grossing, Processing and Embedding
- Cassette design and flow characteristics tested to ensure efficient reagent exchange
- Opening and closing forces adjusted for ease of use, while safeguarding valuable tissue samples
Sectioning
- Precision engineering for solid connection to microtomes
Archiving
- Cassette surface texture optimizes scannability for archiving
Specifications
Yes | |
Small Squares | |
Yes |
750/Cs. | |
Separate Lid, Front Hinge | |
Cassette II Biopsy Cassette |