Learn More
Eppendorf™ Centrifuge 5425/5425 R
Eppendorf’s newest 24-place microcentrifuges, Centrifuge 5425 and 5425 R, feature ergonomic operation and a large selection of rotors with a maximum speed of up to 21,300 x g, making them perfectly suited for all modern molecular biological applications in Eppendorf and PCR tubes from 0.2 to 5 mL.
2420.00 € - 6415.00 €
Product Code | Brand | Model | Refrigerated | Rotor Included | Electrical Requirements | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Model | Refrigerated | Rotor Included | Electrical Requirements | Price | Quantity & Availability | ||
15801645
|
Eppendorf™
5405000263 |
Centrifuge 5425 (EU-IVD), keypad | No | No | 230V/50–60Hz (GB) |
2550.00 €
Each |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
15841645
|
Eppendorf™
5405000468 |
Centrifuge 5425 (EU-IVD), rotary knobs | No | Yes | 230V/50–60Hz (GB) |
3160.00 €
Each |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
15861635
|
Eppendorf™
5405000069 |
Centrifuge 5425 (EU-IVD), keypad | No | Yes | 230V/50–60Hz (GB) |
3160.00 €
Each |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
15881645
|
Eppendorf™
5405000662 |
Centrifuge 5425 (EU-IVD), rotary knobs | No | No | 230V/50–60Hz (GB) |
2550.00 €
Each |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
15801655
|
Eppendorf™
5405000760 |
Centrifuge 5425, rotary knobs | No | No | 230V/50–60Hz (GB) |
2420.00 €
Each |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
15821645
|
Eppendorf™
5405000360 |
Centrifuge 5425, keypad | No | No | 230V/50–60Hz (GB) |
2420.00 €
Each |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
15861645
|
Eppendorf™
5405000565 |
Centrifuge 5425, rotary knobs | No | Yes | 230V/50–60Hz (GB) |
3005.00 €
Each |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
15881635
|
Eppendorf™
5405000166 |
Centrifuge 5425, keypad | No | Yes | 230V/50–60Hz (GB) |
3005.00 €
Each |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
16865100
|
Eppendorf™
5406000160 |
Centrifuge 5425 R, keypad | Yes | Yes | 230V/50–60Hz (GB) |
6110.00 €
1 set |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
16805110
|
Eppendorf™
5406000364 |
Centrifuge 5425 R, keypad | Yes | No | 230V/50–60Hz (GB) |
5685.00 €
Each |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
16845110
|
Eppendorf™
5406000569 |
Centrifuge 5425 R, rotary knobs | Yes | Yes | 230V/50–60Hz (GB) |
6110.00 €
1 set |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
16885110
|
Eppendorf™
5406000763 |
Centrifuge 5425 R, rotary knobs | Yes | No | 230V/50–60Hz (GB) |
5685.00 €
Each |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
16845100
|
Eppendorf™
5406000062 |
Centrifuge 5425 R (EU-IVD), keypad | Yes | Yes | 230V/50–60Hz (GB) |
6415.00 €
1 set |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
16885100
|
Eppendorf™
5406000267 |
Centrifuge 5425 R (EU-IVD), keypad | Yes | No | 230V/50–60Hz (GB) |
5970.00 €
Each |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
16825110
|
Eppendorf™
5406000461 |
Centrifuge 5425 R (EU-IVD), rotary knobs | Yes | Yes | 230V/50–60Hz (GB) |
6415.00 €
1 set |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
16865110
|
Eppendorf™
5406000666 |
Centrifuge 5425 R (EU-IVD), rotary knobs | Yes | No | 230V/50–60Hz (GB) |
5970.00 €
Each |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
16682832
|
Eppendorf™
5405000182 |
Centrifuge 5425, rotary knobs | No | With Rotor FA-18x2-Kit | 230 V, 50/60 Hz |
2875.00 €
Each |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
Description
For processing temperature-sensitive samples, the refrigerated Centrifuge 5425 R provides precise temperature control with its advanced cooling system.- Aerosol-tight Eppendorf QuickLock™ lids for simple and safe handling
- Outstanding software features (e.g. adjustable acceleration and braking ramps, 3 program buttons (only Centrifuge 5425), short function without continuous pressing of the key)
- Extended rotor program (e.g. 5.0 mL rotor, swing-bucket rotor for PCR strips)
- OptiBowl™ design for quiet operation, even without a rotor lid
- Soft-touch one-finger lid closure for ergonomic operation
- Max. rotor capacity: 10 × 5.0 mL vessels
- Max. speed: 21,300 × g (15,060 rpm)
Specifications
21,300 x g |
The Centrifuge 5425 is designed for separating liquid substance mixtures with different densities, in particular, for processing and analyzing samples from the human body in in-vitro diagnostic applications to ensure that the in-vitro diagnostic device can be used according to its intended purpose. This centrifuge including components is an in-vitro diagnostic device according to Directive 98/79/EC of the European Parliament and the Council dated October 27, 1998. Eppendorf centrifuges are exclusively intended for indoor use by trained specialists.