Learn More
Diversey™ ClearKlens Oxifast Sterile Disinfectant
ClearKlens Oxifast is designed for disinfecting hard surfaces and equipments in critical and non-critical areas.
Brand: Diversey™ 100934589
Additional Details : Weight : 8.08000kg
Description
- ClearKlens Oxifast is used for the disinfection of walls, floors, surfaces, and process equipments in the Life Sciences industry.
- When sterile, ClearKlens Oxifast is double bagged for an easy transfer into the cleanroom.
- ClearKlens Oxifast passes EN 13704 test with a shortened contact time of only 5 minutes, an important selection criterion for a Cleanroom disinfectant where wet contact times are limited.
Benefits
- Ready to use sporicidal agent
- Broad spectrum activity even at very short contact time: bactericidal, yeasticidal, fungicidal, virucidal, sporicidal
- Low environmental impact
- Sterile packs are terminally irradiated
- Delivered with certificates of conformity
Specifications
2.3 | |
Ready to use peracetic acid based solution | |
1 L |
Sterile | |
8 Pk. |