missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Clinical Lab Mat

Fisherbrand™ Clinical Lab Mat safely facilitates workflow processes with organization and easy clean up, while keeping benchtops clean and safe from stains or spills.

Brand:  Fisherbrand™

30.93 EUR valid until 2024-04-30
Use promo code "21656" to get your promotional price.



Additional Details : Weight : 0.32000kg

Product Code. 16361648

  • 46.44 € / Each
Estimated Shipment: 06-06-2024
to see stock.
Add to basket

Description

Description

  • Small footprint
  • Accommodates most work spaces
  • Helps facilitate workflow processes, such as spinning blood tubes in centrifuge workstations
  • Gridded surface for tube staging, preventing movement of small objects and securing items such as reservoirs, pipette tip boxes, assay plates, etc
  • Gridded surface enables tubes to be easily marked and identified
  • Personal size is easy to transport and allows users to easily create and protect temporary work areas
  • Reversible
  • Side One: combination gridded and smooth surface
  • Side Two: smooth surface only, two color options available
  • Lip around the edge can hold approx. 250 mL of liquid
Specifications

Specifications

Clinical Lab Mat
12 x 14.9 in. (30.5 x 37.9 cm)
Blue
0.1 in.
200°C
1
Silicone
Silicone
No
392°F
10
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers