Learn More
Epredia™ Cytospin™ 4 Cytocentrifuge Sealed Head Rotor
Use these accessories with the Epredia™ Cytospin™ 4 Cytocentrifuge.
Brand: Epredia™ 59910018
1092.60 EUR valid until 2025-03-30
Use promo code "24134" to get your promotional price.
This item is not returnable.
View return policy
Description
- Shorten preparation time by having a second sealed head ready when the first finishes processing
- Autoclavable
- 12-chamber head seals completely
Specifications
Sealed Head Roto | |
Cytospin 4 Cytocentrifuge | |
Sealed Head Rotor |