missing translation for 'onlineSavingsMsg'
Learn More

IKA™ Digital 2D Rocker

Brand:  IKA™ 0004003000

Code : A0

Additional Details : Weight : 1.50000kg

Product Code. 13480577

  • 1105.00 € / Each
Estimated Shipment: 10-06-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

2.5 to 15°
0 to 80 rpm
4°C to 50°C
1000 mA
50/60 Hz
150 mm
280 mm
2 kg
100/240 V
24 W
DIN EN 60529
330 mm
150 mm
80%
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers