Learn More
3M™ Disposable Particulate Respirator, 8000 Series
Ensures comfortable wear over a range of face sizes. The 3M 8000 Series Particulate Respirator has a convex shape.
25.08 € - 61.25 €
Product Code | Brand | Filter Type | With Exhalation Valve | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Filter Type | With Exhalation Valve | Price | Quantity & Availability | ||||
12302399
|
3M™
7000006980 |
FFP1 NR D | Yes |
30.15 €
Pack of 10 |
In Stock
Log in to see stock. |
||||
12382389
|
3M™
7000034734 |
FFP1 NR D | No |
33.13 €
Pack of 20 |
In Stock
Log in to see stock. |
||||
11713363
|
3M™
7100003010 |
FFP2 NR D | No |
61.25 €
Pack of 20 |
In Stock
Log in to see stock. |
||||
11736003
|
3M™
7100010817 |
FFP2 NR D | Yes |
53.25 €
Pack of 10 |
Estimated Shipment: 17-06-2024
Log in to see stock. |
||||
11733363
|
3M™
7100101091 |
FFP2 | Yes |
32.55 €
Pack of 10 |
In Stock
Log in to see stock. |
||||
11723363
|
3M™
7000006980 |
FFP1 NR D | Yes |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
11703363
|
3M™
7000034734 |
FFP1 NR D | No |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
3M Particulate Respirator P1 P1 for mechanically generated particles grinding, sanding, crushing and sawing Not TGA Approved Not allowed for Clinical Use
The 3M™ Respirators 8000 Series provide lightweight, comfortable and effective respiratory protection against dust and mist. The convex shape, twin strap design, nose foam and nose clip ensure comfortable wear over a range of face sizes. The collapse resistant shell and proprietary 3M™ respirators offer durable, comfortable protection particularly in hot and humid conditions. Dusts, mists and metal fumes are the most common respiratory hazards in the working environment as by-products of processes such as grinding and mixing. 3M respirators provide comfortable, maintenance-free protection against these particular hazards.