Learn More
Dräger™ Short-Term Detector Tubes: Carbon Dioxide
Detector tubes
106.00 € - 271.00 €
Specifications
Components Measured Carbon Dioxide
Quantity 10
View More Specs
Products 5
Product Code Brand Measurement Range Tube Type Humidity Range Price Quantity & Availability  
Product Code Brand Measurement Range Tube Type Humidity Range Price Quantity & Availability  
10629503
View Documents Promotion Details
Dräger™
6728521
100 to 3000 Carbon dioxide 100/a-p 23mg/L
271.00 €
Pack of 10
10251161
View Documents Promotion Details
Dräger™
8101811
100 to 3000 Carbon dioxide 100/a 23mg/L
229.00 €
Pack of 10
10167340
View Documents Promotion Details
Dräger™
CH23501
0.5 to 6% Carbon dioxide 0.1%/a2 30mg/L
106.00 €
Pack of 10
10661923
View Documents Promotion Details
Dräger™
CH31401
0.5 to 10% Carbon dioxide 0.5%/a 50mg/L
148.00 €
Pack of 10
10113112
View Documents Promotion Details
Dräger™
CH20301
5 to 60% Carbon dioxide 5%/A 50mg/L
109.00 €
Pack of 10
Description
Description
Detector tubes
Glass detector tubes are filled with an inert reagent carrier material (silica gel, aluminum oxide, or aluminum silicate) impregnated with an indicating reagent. Reagent produces a colorimetric indication in the presence of a particular gas, vapor, or aerosol. Concentrations can be read directly from the color change on the tube's printed scale. Tubes are hermetically sealed. Most have a two-year shelf life (when stored at room temperature). Available in packs of ten.
Note: Some tests require a pretube or comparison tube in addition to the detector tube. These tubes are included in the box of 10, thus reducing the number of detector tubes. The actual number of detector tubes is cited in the ordering charts.
Order Info
Extension of measuring range listed is possible for some detector tubes. Call Fisher Scientific™ at 1-800-772-6733 for more information.
Specifications
Specifications
Carbon Dioxide
10