Learn More
Invitrogen™ EVOS™ M5000 Stage Plate
This EVOS M5000 Stage Plate accommodates a wide range of multi-well plates and all EVOS vessel holders designed for a mechanical stage.
Brand:  Invitrogen™ AMEPVH089
Additional Details : Weight : 0.68000kg
Product Code. 16376602
  • 307.00 € / Each
Estimated Shipment: 24-06-2024
to see stock.
Add to basket
Description
Description
Individual vessel holders are readily switched, making it easy to image samples in a wide range of vessel formats. Vessel holders are available for microscope slides, cell culture flasks, Petri dishes, microwell plates, and more.
The EVOS M5000 Stage Plate offers these advantages:
• Perfect fit of multi-well plates
• Compatible adapters for a wide-range of vessel holders
• Plates and holders are easily changed
Specifications
Specifications
EVOS M5000
Stage Plate
Room Temperature
EVOS
Cellular Imaging