Learn More
SCAT™ Funnels and Adapters for Barrels and Drums with Mechanical Level Control
Robust SCAT™ 'ARNOLD' Safety Funnels with adapters and mechanical level controls for use on various drums with different thread types. Also available in electrically dissipative variants for the safe disposal of explosive liquid waste.
243.00 € - 471.00 €
Product Code | Brand | Closure Type | Includes | Material | Product Type | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Closure Type | Includes | Material | Product Type | Price | Quantity & Availability | ||
17893510
|
SCAT™
107889 |
S60 (f) to GL45 (m) | ARNOLD Funnel Adapter, Mechanical Level Control | HDPE-EC | Funnel Adapter |
316.00 €
Each |
Estimated Shipment: 20-06-2024
Log in to see stock. |
||
17838733
|
SCAT™
317631 |
R2 BSP/G2 (m) and BCS 56 x 4 (m) to GL45 (m) | ARNOLD Funnel, Lid, Splash Guard, Removable Sieve, Grounding Cable | HDPE-EC | Safety Funnel |
243.00 €
Each |
Estimated Shipment: 18-06-2024
Log in to see stock. |
||
17848743
|
SCAT™
317650 |
R2 BSP/G2 (m) and BCS 56 x 4 (m) to GL45 (m) | ARNOLD Funnel, Lid, Splash Guard, Removable Sieve, Grounding Cable, Funnel Adapter, Mechanical Level Control | HDPE-EC | Safety Funnel |
471.00 €
Each |
Estimated Shipment: 18-06-2024
Log in to see stock. |
||
17868743
|
SCAT™
317660 |
R2 BSP/G2 (m) and BCS 56 x 4 (m) to GL45 (m) | ARNOLD Funnel, Lid, Splash Guard, 2 Removable Sieves, Funnel Adapter, Mechanical Level Control | HDPE | Safety Funnel |
422.00 €
Each |
Estimated Shipment: 18-06-2024
Log in to see stock. |
||
Description
- Safety funnels based on the SCAT™ 'ARNOLD' funnel series
- Ready-to-use sets consisting of safety funnels and barrel adapters
- Made of high-quality plastics for durable and long-lasting use
- Adapters can be combined with all GL45 safety funnels of the 'ARNOLD' and 'MARCO' series
- Available for all common thread types and sizes
- Also available in electrically conductive material for hazardous areas
Specifications
SCAT ARNOLD Funnel |