missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gilson™ PIPETMAN™ M Pipettes
Single or multichannel. Equipped with motorized piston that virtually eliminates pipetting variability among users and minimizes RSI (Repetitive Strain Injuries) among users
469.00 € - 980.00 €
Specifications
Product Line | Pipetman™ |
---|---|
Type | Pipette |
Finger Style | Ergonomic |
Warranty | 1 Year |
Product Code | Brand | Number of Channels | Model | Volume (Metric) | Compatible Tips | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Number of Channels | Model | Volume (Metric) | Compatible Tips | Price | Quantity & Availability | ||
15560500
|
Gilson™
F81025 |
8 | P8x10M | 0.5 to 10 μL | D10, DL10, DF10, DFL10 tips | N/A | |||
15570500
|
Gilson™
F81026 |
12 | P12X10M | 0.5 to 10 μL | D10, DL10, DF10 or DFL10 tips | N/A | |||
15580500
|
Gilson™
F81027 |
8 | P8X20M | 1 to 20 μL | DL10, DF30ST, D200 tips | N/A | |||
15590500
|
Gilson™
F81028 |
12 | P12x20M | 1 to 20 μL | DL10, DF30ST, D200 tips | N/A | |||
15500510
|
Gilson™
F81029 |
8 | P8x200M | 20 to 200 μL | D200, D300, DF100ST, DF200ST, DF300ST tips | N/A | |||
15510510
|
Gilson™
F81030 |
12 | P12x200M | 20 to 200 μL | D200, D300, DF100ST, DF200ST, DF300ST tips | N/A | |||
15520510
|
Gilson™
F81031 |
8 | P8x300M | 10 to 300 μL | D10, DL10, DF10, DFL10 tips | N/A | |||
15530510
|
Gilson™
F81032 |
12 | P12x300M | 10 to 300 μL | D10, DL10, DF10 or DFL10 tips | N/A | |||
15550490
|
Gilson™
F81014 |
1 | P300M | 20 to 300 μL | D200, DF200, D300, DF300 | N/A | |||
15560490
|
Gilson™
F81015 |
1 | P1200M | 100 to 1200 μL | D1000, DF1000, D1200, DF1200 | N/A | |||
15570490
|
Gilson™
F81016 |
1 | P5000M | 500 to 5000 μL | D5000 | N/A | |||
15580490
|
Gilson™
F81017 |
1 | P10mLM | 1 to 10 mL | D10 mL | N/A | |||
15590490
|
Gilson™
F81018 |
8 | P8x100M | 10 to 10 μL | D200 | N/A | |||
15500500
|
Gilson™
F81019 |
12 | P12x100M | 10 to 100 μL | DF100 | N/A | |||
15510500
|
Gilson™
F81020 |
8 | P8x1200M | 50 to 1200 μL | D1200 | N/A | |||
15520500
|
Gilson™
F81021 |
12 | P12x1200M | 50 to 1200 μL | DF1200 | N/A | |||
Description
- Motorized piston with adjustable piston speed that is compatible with varying solution densities
- Lockable volume to prevent inaccuracies
- Color-coded push-button corresponds to matching tip packs
- Service alert reminders
- Unique tip holder design allows for a secure fit of most brands of tips
- Features four essential pipetting modes dash (en) reverse, mix, repetitive and pipetting to best fit your application
- Pipette can be used while charging eliminating bottlenecks and downtime
- Designed to exert equal pressure on each piston to ensure consistent volume levels across all channels
- Power carrousel conveniently holds and charges up to 5 PIPETMAN M pipettes at once, charging 80% of its full capacity in less than one hour
- Gilson Trident and unique spacer distribute ejection forces evenly over each tip, reducing the force required to eject tips
Specifications
Pipetman™ | |
Pipette | |
Ergonomic | |
1 Year |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title