missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Heating/Cooling Mini Shakers

Ideal for analyses that requires a stable, controlled temperature.

Brand:  Fisherbrand™

Additional Details : Weight : 21.15000kg

 View more versions of this product

Product Code. 15544080

  • 2520.00 € / Each
Estimated Shipment: 04-07-2024
to see stock.
Add to basket

Description

Description

  • Microprocessor control -The variable speed microprocessor control provides consistent, uniform shaking action. Microprocessor will display last set-point and will restart if power is interrupted.
  • PID temperature controller - Maintains precise temperature control from 10°C below ambient to 65°C. Easy-to-use controls allow users to adjust temperature in 1°C increments.
  • Triple eccentric drive - Permanently lubricated ball bearings and maintenance free, brushless DC motor provide reliable service and continuous duty operation.
  • LED display- Touch pad controls with easy-to-read, independent LED displays for temperature, speed and time allow operator to view all settings at once. Provides repeatable and accurate results every time and is easily visible across lab benches. Timer will display elapsed time or, when programmed to user defined limit, will shut off unit when time reaches zero. Display will show last used settings, even after power has been turned off.
  • Temperature calibration mode - Allows user to calibrate unit to an external temperature device.
  • Applications - Cell and bacterial cultures, hybridizations, and enzyme reactions
Specifications

Specifications

3 mm
150 to 1200 rpm (Microplate), 100 to 600 rpm (Modular Block)
EU, UK, CN Plug
16.3 x 11.5 x 12 in. (415 x 293 x 308 mm)
LED
1 sec. to 9999 min.
±2% of set speed up to 999 rpm ±5% 1000 to 1200 rpm
230 V,1 A, 450 W
7 x 5.5 in. (180 x 140 mm)
Audible and Visual
Maximum 80%
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers