Learn More
SCAT™ HPLC Waste Sets, V2.0
HPLC Waste Sets, V2.0 are economical combinations for clean and safe disposal of liquid laboratory waste by HPLC systems or other analytical instruments. All sets consist of a canister and an original SCAT™ SafetyCap with exhaust filter as well as fittings, blind plugs and a tube connector.
Brand:  SCAT™ 307367
175.76 EUR valid until 2024-12-31
Use promo code "23290" to get your promotional price.
Additional Details : Weight : 0.35000kg
 View more versions of this product
Product Code. 17845714
  • 239.00 € / 1 set
Estimated Shipment: 02-07-2024
to see stock.
Add to basket
Description
Description
SCAT™ offers a complete system for safe supply and disposal of solvents in analytics. All products are compatible across generations and perfectly matched to each other.
  • Customized system with high-quality set components
  • Safety canisters made of HDPE or polypropylene material
  • Attractive price compared to single purchase
  • Ready to use, no further components necessary
Specifications
Specifications
Safety Waste Cap, 2 x PFA fitting 2.3 mm OD, 2 x PFA fitting 3.2 mm OD, 1 x PP tube connector 5 to 11.5 mm ID, 1 x GL14 connection for exhaust filter, 1 x canister, S55, 5 L, PE-HD, 1 x exhaust filter S, with splash guard and change indicator
HPLC Systems
S55
5 L
Set 6