missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ FetalClone™ I Serum (U.S.)
Optimized for growth of hybridomas, supplement cell culture growth with U.S.-sourced HyClone™ FetalClone™ I Serum, a fetal bovine serum (FBS) alternative containing comparable IgG levels to those found in FBS.
91.70 € - 382.00 €
Specifications
Certifications/Compliance | Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities |
---|---|
Country of Origin | United States |
Product Type | FBS Replacement Serum |
Sterility | Sterile |
Sterilization Method | Sequential-filtered, 100 nm |
Product Code | Brand | Quantity | Heat Inactivated | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Heat Inactivated | Price | Quantity & Availability | ||||
10275022
|
Cytiva
SH30080.02 |
100 mL | No |
91.70 €
100mL |
Estimated Shipment: 25-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10780245
|
Cytiva
SH30080.03 |
500 mL | No |
382.00 €
500mL |
Estimated Shipment: 26-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12329832
|
Cytiva
SH30080.02HI |
100 mL | Yes |
111.00 €
100mL |
Estimated Shipment: 16-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
Specifications
Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities | |
FBS Replacement Serum | |
Sequential-filtered, 100 nm | |
≤ 20 mg/dL | |
USDA Inspected |
United States | |
Sterile | |
≤ 10 EU/mL | |
No |