missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydroxynaphthol Blue, Indicator for Metal Titration, Honeywell Fluka™
98.44 EUR valid until 2024-05-31
Use promo code "21664" to get your promotional price.
Use promo code "21664" to get your promotional price.
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass Bottle
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
UN Number | NONH for all modes of transport |
Infrared Spectrum | Conforms to structure |
Formula Weight | 620.47g/mol |
Packaging | Glass Bottle |
Quantity | 10 g |
Physical Form | Liquid |
Chemical Name or Material | Hydroxynaphthol blue |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'