missing translation for 'onlineSavingsMsg'
Learn More

Memmert™ CO2 Incubator ICO

Ideal for Incubation with CO2, Biosyntesis and Cell Incubation in the Laboratory.

Brand:  Memmert™ ICO240


Code : NEW

Additional Details : Weight : 110.00000kg

 View more versions of this product

Product Code. 15692244

  • 9186.00 € / Each
Estimated Shipment: 01-10-2024
to see stock.
Add to basket

Description

Description

Sterile, safe and reliable – these three words describe the Memmert CO2 incubator ICO for industrial use.

  • Model sizes: 50, 105, 150, 240
  • Temperature adjustment range: +18 °C to +50 °C
  • CO2 adjustment range: 0 to 20 %
  • Humidity adjustment ranges: 40 to 97 % rh (with option K7)
  • O2 adjustment ranges: 1 to 20 % (with option T6)
  • 1 model variant: TwinDISPLAY (intuitive control)
  • AtmoCONTROL software on a USB stick for programming, managing and transferring programmes via Ethernet interface or USB port
  • Perfect distribution of temperature, humidity and CO2
  • Long-time stable and repetitively accurate
  • The interior chamber, including all installations and sensors, can be sterilised at +180 °C in a 60-minute programme
  • Visual and acoustic alarm as well as alarm notification to mobile phones if the individually adjustable temperature, CO2, O2, or humidity ranges are exceeded
  • Battery-buffered ControlCOCKPIT: The operating display, logging and CO2 control are fully functional even during a power failure
  • Fail-safe, FDA-compliant logging of temperature, CO2, O2 humidity, open door time data
  • Adjustable parameters: temperature (Celsius or Fahrenheit), CO2, programme time, time zones, summertime/wintertime

Active humidity control ensures short recovery times after the door has been opened and minimises vaporisation in the interior. Together with the heating of the interior from all six sides and the heated inner glass door, it offers maximum protection for cell and tissue cultures and avoids the dangerous formation of condensation. The turbulence-free chamber ventilation ensures a constant and uniform atmosphere.

Specifications

Specifications

Optic and acoustic
TwinDISPLAY: adaptive multifunctional digital PID-microprocessor controller with 2 high-definition TFT-colour displays
Stainless Steel
IR Sensor
2
40 to 97%
CO2 Incubator
2 high-definition TFT-colour displays (TwinDISPLAY)
Digital backwards counter with target time setting, adjustable from 1 minute to 99 days
691 x 759 x 1176 mm
110 kg
115/230 V
15 kg
241 L
0 to 20%
Textured Stainless Steel, Rear Zinc-plated Steel
1 to 20%
18°C to 50°C
60 min.
Perforated stainless steel
500 x 600 x 810 mm
145 kg
115/230 V, 50/60 Hz
50/60 Hz
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers