Learn More
Eppendorf™ Universal Platform for the Innova™ S44i Incubator Shaker
A selection of universal and dedicated platforms for Erlenmeyer, Fernbach or Lauber Thomson flasks.
Brand:  Eppendorf™ S44I040001
1387.36 EUR valid until 2024-12-31
Use promo code "23275" to get your promotional price.
Additional Details : Weight : 10.20000kg
Product Code. 15888361
  • 1460.00 € / Each
In Stock
to see stock.
Add to basket
Description
Description
  • 9 sticky pads necessary to cover platform
  • Sticky pads sold separately
  • Anodized aluminum
Specifications
Specifications
Aluminum
Innova™ S44i
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products
Special Offers
Special Offers
Services