missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ Lens Cleaning Solution
46.15 EUR valid until 2024-12-31
Use promo code "22912" to get your promotional price.
46.15 EUR valid until 2024-06-30
Use promo code "21674" to get your promotional price.
Use promo code "22912" to get your promotional price.
46.15 EUR valid until 2024-06-30
Use promo code "21674" to get your promotional price.
Click to view available options
Size: 120 mL
Description
- Available in 120 mL bottle with pump action spray
- Effective cleaning solutions for a range of work environments
Specifications
Product Type | Lens Cleaning Solution |
For Use With (Equipment) | Lens |
Includes | Pump action spray |
Quantity | 120 mL |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'