Learn More
Cytiva (formerly Pall Lab) Lid Kit for Funnel
Replacement lid kit for funnels with unique magnetic seal.
Brand:  Cytiva (formerly Pall Lab) 4246
Additional Details : Weight : 1.03550kg
Product Code. 17174281
  • 96.25 € / Each
Estimated Shipment: 28-06-2024
to see stock.
Add to basket
Description
Description
  • Unique magnetic seal allows easy, one-handed vacuum filtration of liquids with compatible membranes
  • Not approved for use in medical, clinical, surgical or other patient applications
Specifications
Specifications
300 mL Funnel
Polyphenylsulfone Lid, Polypropylene Vent Plug
Non-sterile
Silicone O-ring