missing translation for 'onlineSavingsMsg'
Learn More

Attune™ NxT Flow Cytometer, blue/violet

Attune NxT Flow Cytometer, blue/violet

Brand:  Invitrogen A24862


Additional Details : Weight : 26.00000kg

Product Code. 15300677

  • POA / Each
Estimated Shipment: 17-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.


Description

Description

The Attune™ NxT Acoustic Focusing Cytometer is an easy-to-use instrument designed to detect cell size and granularity, as well as fluorescence expression of labeled antibodies, proteins, and functional reagents. The innovative design of the instrument results in precise data and acquisition speeds up to 10x faster than other flow cytometers. The design also eliminates lengthy sample preparation procedures, reduces the time to analyze a file, allows small or limited samples to be analyzed, and minimizes variability in measurements, leading to better resolution of populations. It is ideal for immunophenotyping and signaling studies, cell cycle analysis, detection of rare events, stem cell analysis, cancer and apoptosis studies, and microbiological assays.

With the Attune™ NxT Acoustic Focusing Cytometer you get:
  • A flexible platform that provides highly sensitive and accurate measurement of cell sizeand granularity, as well as fluorescence detection
  • State of the art optics for long-term reliability
  • Acoustic focusing for high-precision data
  • An intuitive software user interface that reduces time to proficiency
  • Improved workflow to reduce sample preparation time before analysis
  • Faster data analysis that leads to reduced acquisition times

    Flexible design that accommodates changing needs, budget, or laboratory space
    The instrument’s modular design accommodates the most common fluorophores and fluorescent proteins used in flow cytometry enabling alignment with previous experimental results and accommodating future needs through simple upgrades. The small footprint of the instrument requires minimal bench space, even allowing use in a cell culture hood.

    Acoustic focusing improves efficiencies compared to traditional flow cytometers
    Traditional flow cytometers rely on a large volume of fluid to focus cells prior to analysis, resulting in extensive sample preparation prior to analysis and introducing the possibility of error due to artifacts created during the sample preparation process. With the Attune™ NxT Acoustic Focusing Cytometer, blood samples can be run without being manipulated using the No-Wash No-Lyse blood cell assay. The instrument also offers flexibility in the sample concentration that can be analyzed.

    Acoustic focusing enables:
  • Elimination of concentration steps
  • Effective background fluorescence reduction by a simple dilution step, eliminating the need to wash fluorescence samples before analysis
  • No-lyse/no-wash assays
  • Analysis of samples that have low cell numbers, including CSF, stem cells, mouse bone marrow, mouse blood, and thin needle aspirates
  • Acquisition of a high volume of samples in minimal time

    Learn more about the Attune NxT cytometer and see all available configurations >
    Learn more about how acoustic focusing works >

  • Specifications

    Specifications

    • Attune NxT Flow Cytometer
    • Minitower Windows 10 64-bit computer with Attune Cytometric Software
    • Power cord with adapters
    • USB cable
    • Attune Shutdown Solution
    • Attune Wash Solution
    • Attune Focusing Fluid
    • Attune NxT Flow Cell Cleaning Solution
    • Attune Performance Tracking Beads
    • Quick reference card

    Tube(s)
    50/60 Hz
    Attune™ NxT Acoustic Focusing Cytometer
    Less hazardous
    Blue Excitation Laser (488 nm), Violet Excitation Laser (405 nm)
    Attune™
    Room Temperature
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Certificates
    Special Offers

    Special Offers

    For Research Use Only. Not for use in diagnostic procedures.