Learn More
Invitrogen™ Anza™ 10-Pack Starter Kit
Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.
Brand:  Invitrogen™ IVGN3006
Additional Details : Weight : 0.50000kg
Product Code. 15441933
  • 390.00 € / 1 set
Estimated Shipment: 07-06-2024
to see stock.
Includes: Anza™ 11 EcoRI, Anza™ 5 BamHI, Anza™ 1 NotI, Anza™ 16 HindIII, Anza™ 12 XbaI, Anza™ 3 BcuI, Anza™ 8 XhoI, Anza™ 10 DpnI, Anza™ 19 BglII, Anza™ 26 Eco32I, Anza™ T4 DNA Ligase Master Mix, Anz
Description
Description
Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.
The Invitrogen Anza Restriction Enzyme Cloning System utilizes a single buffer system for complete digestions and incorporates Anza cloning enzymes into a clear and simple system.
  • One buffer for all restriction enzymes
  • One digestion protocol for all DNA substrates
  • Complete digestion in 15 min
  • Includes 10 common restriction enzymes
  • Complete with Anza T4 DNA Ligase Master Mix and Anza Alkaline Phosphatase
Certifications
CoA
Specifications
Specifications
20 μL each of Anza 11 EcoRI, Anza 5 BamHI, Anza 1 NotI, Anza 16 HindIII, Anza 12 XbaI
20 μL each of Anza 3 BcuI, Anza 8 XhoI, Anza 10 DpnI, Anza 19 BglII, Anza 26 Eco32I
100 μL Anza T4 DNA Ligase Master Mix
20 μL Anza Alkaline Phosphatase
500 μL Anza Buffer (10X)
500 μL Anza Red Buffer (10X)
Store at -5 to -30°C.
Methylation Sensitivities Varies by Enzyme
Anza™ 10X Red Buffer, Anza™ 10X Buffer
Starter Kit
Anza™
Approved for shipment on Wet or Dry Ice
HindIII, NotI, Alkaline Phosphatase, XhoI, BcuI, XbaI, DpnI, T4 DNA Ligase, Eco32I, BamHI, BglII, EcoRI
No
Traditional Cloning