Learn More
SCAT™ Safety Funnel 'MARCO' with Lid, V2.0
The 'MARCO' safety funnel can be safely closed and prevent danger of ignition, and is also available in conductive HDPE-EC material.
108.05 € - 203.00 €
Specifications
Diameter (Metric)Top | 140 mm |
---|---|
Capacity (Metric) | 300 mL |
Material | HDPE |
Product Type | Safety Funnel |
Product Code | Brand | Height (Metric) | Includes | Stopper Number | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Height (Metric) | Includes | Stopper Number | Price | Quantity & Availability | |||
16832140
|
SCAT™
318992 |
370 mm | Funnel with Hinged Lid and Lance | GL45 |
125.00 €
Each |
Estimated Shipment: 14-06-2024
Log in to see stock. |
|||
16609095
|
SCAT™
318952 |
150 mm | Funnel with Hinged Lid, No Lance | GL45 |
112.00 €
Each |
In Stock
Log in to see stock. |
|||
16862140
|
SCAT™
318995 |
370 mm | Funnel with Hinged Lid and Lance | S50 |
125.00 €
Each |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||
16629095
|
SCAT™
318955 |
150 mm | Funnel with Hinged Lid, No Lance | S50 |
112.00 €
Each |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||
16822130
|
SCAT™
318953 |
150 mm | Funnel with Hinged Lid, No Lance | S51 |
112.00 €
Each |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||
16842140
|
SCAT™
318993 |
370 mm | Funnel with Hinged Lid and Lance | S51 |
125.00 €
Each |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||
16822140
|
SCAT™
318991 |
370 mm | Funnel with Hinged Lid and Lance | S55 |
125.00 €
Each |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||
16699085
|
SCAT™
318951 |
150 mm | Funnel with Hinged Lid, No Lance | S55 |
115.58 €
Each |
In Stock
Log in to see stock. |
|||
16812140
|
SCAT™
318990 |
370 mm | Funnel with Hinged Lid and Lance | S60/61 |
125.00 €
Each |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||
16689085
|
SCAT™
318950 |
150 mm | Funnel with Hinged Lid, No Lance | S60/61 |
112.00 €
Each |
In Stock
Log in to see stock. |
|||
16852140
|
SCAT™
318994 |
370 mm | Funnel with Hinged Lid and Lance | S65 |
125.00 €
Each |
Estimated Shipment: 21-06-2024
Log in to see stock. |
|||
16619095
|
SCAT™
318954 |
150 mm | Funnel with Hinged Lid, No Lance | S65 |
112.00 €
Each |
Estimated Shipment: 21-06-2024
Log in to see stock. |
|||
17898723
|
SCAT™
318957 |
370 mm | Funnel with Hinged Lid, No Lance | CPC/Justrite |
203.00 €
Each |
Estimated Shipment: 19-06-2024
Log in to see stock. |
|||
17957260
|
SCAT™
318956 |
150 mm | Funnel with Hinged Lid, No Lance | S70/71 |
108.05 €
Each |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17947250
|
SCAT™
318996 |
370 mm | Funnel with Hinged Lid and Lance | S70/71 |
120.78 €
Each |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
- Modern lid funnel made of high-quality HDPE with very high resistance to acids, alkalis and many chemicals
- Integrated gripping edge on the funnel lid increases the grip without having to touch the possibly contoured lid
- Innovative funnel lid with integrated sealing ring closes the funnel edge and prevents the escape of explosive or health-endangering gases
- Sturdy hinge guarantees a long service life of the closing mechanism
- Rotating screw cap makes it easier to screw the lid funnel on and off the canister
- Removable sieve for easy cleaning
- Non-conductive version of the MARCO V2.0 is in line with the SCAT color scheme due to its blue and white coloring
- Conductive versions of the MARCO V2.0 are in black and thus clearly differ in color from the non-conductive version
Spécification
140 mm | |
300 mL | |
HDPE | |
Safety Funnel |