missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Microcentrifuges, Micro 17/17R
Adjustable 99 minute timer and quick-spin feature for short runs.
2835.00 € - 5525.00 €
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
11516873
|
Fisherbrand™
|
Microcentrifuge Micro 17, ventilated |
2835.00 €
Each |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11526873
|
Fisherbrand™
|
Microcentrifuge Micro 17R, refrigerated |
5525.00 €
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Capacity: 48mL (24 x 1.5/2mL tubes)
- Maximum speed/RCF: 13300rpm/17000xg
- Adjustable 99 minute timer and quick-spin feature for short runs
- Supplied with click seal rotor (24 place 1.5/2mL) and biocontainment lid
Specifications
24 x 1.5/2 mL | |
13,300 rpm | |
17,000 x g |