missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Nunc™ MicroWell™ 96-Well, Nunclon Delta-Treated, Flat-Bottom Microplate
116.50 EUR valid until 2024-06-28
Use promo code "21649" to get your promotional price.
116.50 EUR valid until 2024-06-30
Use promo code "21614" to get your promotional price.
155.53 EUR valid until 2024-06-30
Use promo code "21742" to get your promotional price.
155.53 EUR valid until 2024-12-31
Use promo code "21940" to get your promotional price.
Use promo code "21649" to get your promotional price.
116.50 EUR valid until 2024-06-30
Use promo code "21614" to get your promotional price.
155.53 EUR valid until 2024-06-30
Use promo code "21742" to get your promotional price.
155.53 EUR valid until 2024-12-31
Use promo code "21940" to get your promotional price.
Click to view available options
1/Pk., 50/Cs.
10/Pk., 160/Cs.
Description
Thermo Scientific™ Nunc MicroWell 96-Well Microplates are optimized for robotics, automated readers and liquid handling systems. 96-well microplates are used for antibiotic screens, cell-based assays and screening compounds. The flat bottom shape is ideal for microscopic and optical measurements. Our plates are high quality for consistent and reliable results.
- Clear polystyrene has excellent optical properties making it ideal for optical measurements
- Untreated polystyrene is hydrophobic and is suited for growth of suspension cultures that can proliferate and grow without attachment
- Untreated surfaces are ideal for adding coatings in your lab
- Cell Culture Treated: The Nunclon™ Delta cell culture treated surface is a hydrophilic surface that promotes cell attachment and growth
- Plates are optimized for robotics, plate readers and liquid handlers
- Customized bar coding is available
Specifications
Barcode | Custom Barcoding Available |
Certifications/Compliance | All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004) |
Lid | With Lid |
Description | 96F well plate, Nunclon Delta, clear, with lid, Sterile (Pack of 1, Case of 50) |
Array | 8 x 12 |
Packaging | 1/Pk., 50/Cs. |
Product Line | Nunc |
Color | Clear |
Product Type | Microplate |
Material | Polystyrene |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'