missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nunc™ Multidishes with Nunclon™ Supra Surface

Experience optimal performance with Thermo Scientific Nunc plates with Nunclon Supra surface treatment for culturing a wide variety of fastidious adherent cells.

304.00 € - 363.00 €

Specifications

Lid With Lid
Product Line Nunc
Color Clear
Disposable Single-use
Material Polystyrene
View More Specs

Products 4
Product Code Brand No. of Wells Price Quantity & Availability  
Product Code Brand No. of Wells Price Quantity & Availability  
17907472
View Documents Promotion Details
Thermo Scientific™
140680
6
304.00 €
Pack of 75
View alternative products
Save Up to 
»
Estimated Shipment: 21-08-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
17937422
View Documents Promotion Details
Thermo Scientific™
140681
12
316.00 €
Pack of 75
View alternative products
Save Up to 
»
In Stock
to see stock.
Please to purchase this item. Need a web account? Register with us today!
17997422
View Documents Promotion Details
Thermo Scientific™
140682
24
319.00 €
Pack of 75
View alternative products
Save Up to 
»
In Stock
to see stock.
Please to purchase this item. Need a web account? Register with us today!
17957422
View Documents Promotion Details
Thermo Scientific™
140683
48
363.00 €
Pack of 75
View alternative products
Save Up to 
»
Estimated Shipment: 21-08-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Experience optimal performance with Thermo Scientific Nunc plates with Nunclon Supra surface treatment for culturing a wide variety of fastidious adherent cells. Intended for general cell culture applications, the Nunclon Supra surface is more hydrophilic than the Nunclon Delta surface, enhancing the growth of finicky cell types under no-serum or low-serum conditions.

Thermo Scientific Nunc vessels with Nunclon Supra surface treatment are designed for culturing fastidious adherent cells. Intended for general cell culture applications, the Nunclon Supra surface is more hydrophilic than the Nunclon Delta surface, enhancing the growth of finicky cell types under no-serum or low-serum conditions.

The Nunclon Supra surface supports serum-free and coating-free culture of human mesenchymal stromal cells (MSCs), retaining their morphology and promoting high cell viability. Nunclon Supra cultureware is a great xeno-free solution for cell therapy research applications.

Features of Nunc Multidishes with Nunclon Supra Surface include:
• Consistent lot-to-lot performance
• Sterile with SAL 10-6
• Non-cytotoxic
• Non-pyrogenic
• Biocompatible per USP Class VI for raw materials
• Shelf life: 5 years

Specifications

Specifications

With Lid
Clear
Polystyrene
Nunclon Supra
75 Dishes
Nunc
Single-use
Sterile
Flat Bottom (F-well)
1 Dish
Videos
SDS
Documents

Documents

Product Certifications

For Laboratory Use