missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PBS, Phosphate Buffered Saline, 10X Powder, pH 7.4, Fisher BioReagents
Brand: Fisher Bioreagents BP665-1
36.15 EUR valid until 2024-06-30
Use promo code "21742" to get your promotional price.
Code : 32
Additional Details : Weight : 0.21244kg
Description
- Provides a convenient way to make 1L of 10X stock PBS solution
- Eliminates the need to weigh and mix individual components
- When reconstituted in ultrapure water such as BP2819, each packet yields 1L of a 10X PBS solution containing 1.37M NaCl, 0.027M KCl, and 0.119M phosphates
- Tested for the absence of DNase, RNase, and protease
- PBS is a standard buffer used in many biochemical applications to maintain pH
- Intended for laboratory research use only
Specifications
White | |
Foil Pack | |
Solid |
Room temperature | |
2 x 1L |