missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phthalic Anhydride, 99%, Thermo Scientific Chemicals
18.42 EUR valid until 2024-06-30
Use promo code "21742" to get your promotional price.
Use promo code "21742" to get your promotional price.
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
10 kg
Packaging:
Plastic bottle
Plastic drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 129.0°C to 132.0°C |
Boiling Point | 284.0°C |
Flash Point | 152°C |
Assay Percent Range | 99% |
Packaging | Plastic bottle |
Quantity | 250 g |
Beilstein | 17,469 |
Fieser | 01,882; 17,148 |
Merck Index | 15,7484 |
Solubility Information | Solubility in water: 6g/L (20°C). Other solubilities: soluble in alcohol and carbon disulfide, slightly soluble in ether, 7.5% in benzene (25°C), forms phthalic acid in water |
Show More |
Safety and Handling
Product Identifier
- Phthalic anhydride
- Danger
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 1
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
- Specific target organ toxicity Category 3
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H335-May cause respiratory irritation.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'