missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ NHS (N-hydroxysuccinimide)

Prepare amine-reactive NHS-ester crosslinkers or enhance carbodiimide (EDC) conjugation for peptides or proteins using N-hydroxysuccinimide, CAS 6066-82-6.

Brand:  Thermo Scientific™ 24500

Product Code. 10772155

  • 71.25 € / 25g

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Thermo Scientific™ Pierce NHS is pure N-hydroxysuccinimide, the essential compound needed to prepare amine-reactive NHS-ester crosslinkers or to enhance conjugation efficiencies when using the carbodiimide EDC.

Highlights:

  • Form an acyl amino ester with carboxyl groups when used the EDC
  • NHS-activated molecule reacts efficiently with primary amines, resulting in stable amide bond
  • Allows for two-step EDC reactions between carboxyl- and amine-containing molecules
  • NHS-activated molecule can be stabilized by drying, allowing manufacture of amine-reactive probes and surfaces
  • Reactive groups: NHS
  • Reactive toward: amino groups (upon activation of carboxylates)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Amine-Carboxyl
NHS
No
Not determined
Pierce™
Heterobifunctional
Short (<10 Å)
25 g
Standard
Crosslinker
No
115.09
Upon receipt store at 4°C.
Ambient
Chemical Labeling
Carbodiimide
Powder
Water
Yes
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ NHS (N-hydroxysuccinimide) > 25g

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.