Learn More
Thermo Scientific™ Pierce™ Dimethylformamide (DMF), Sequencing grade
Perform liquid chromatography (HPLC) and mass spectrometry (LC-MS) applications with this 99.5%-pure DMF solvent (MW 73.09, CAS 68-12-2) with low UV-absorptivity.
Brand: Thermo Scientific™ 20673
Code : A0
Additional Details : CAS Number : 68-12-2 Weight : 0.05000kg Transport : UN number : 2265 Chem class : 3 Pack group : III
Description
Thermo Scientific Pierce Dimethylformamide (DMF) is a sequencing-grade preparation with high purity and consistent absorptivity properties for use in HPLC and mass spectrometry methods.
This sequencing-grade dimethylformamide is specially purified by a proprietary method and tested to ensure lot-to-lot consistency with a low UV absorbance to provide you with the most sensitive detection across all wavelengths. DMF is packaged in solvent-rinsed amber glass bottles and sealed with PTFE-lined fluorocarbon caps for ultimate protection.
Highlights:
- Highly purified and tested to ensure reliable performance in HPLC, mass spectrometry and sequencing applications
- Packaged in solvent-rinsed amber glass bottles and sealed with PTFE-lined fluorocarbon caps
Product Details:
- General properties of DMF
- Alternative names: n,n-Dimethylformamide, n-Formyldimethylamine
- Molecular formula: C3H7ON
- Molecular weight: 73.09
- Density:0.945g/mL
- CAS number: 68-12-2
Pierce sequencing-grade DMF is specially purified and tested to the highest specifications to ensure the integrity of your data, maximize sensitivity in your assay and prolong the life of your equipment.
Specifications
Dimethylformamide (DMF) | |
Liquid | |
Pierce™ |
Store at room temperature. | |
Solvent (Pure) | |
HPLC, Mass Spectrometry |
Safety and Handling
- Dimethylformamide
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Reproductive toxicity Category 1B
Hazard Statement
- H312-Harmful in contact with skin.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H360D-May damage the unborn child.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : N,N-dimethylformamide
For Research Use Only. Not for use in diagnostic procedures.