missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Polypropylene Clear Autoclave Bags
Designed to be strong, pliable, lean and puncture resistant. Fisherbrand™ Polypropylene Clear Autoclave Bags withstand high temperatures for autoclaving.
27.24 € - 69.96 €
Specifications
Autoclavable | Autoclavable |
---|---|
Material | Polypropylene |
Thickness (Metric) | 40 μm |
Quantity | 100/Pk. |
Temperature Range (Metric) | 134°C to 140°C |
Product Code | Brand | Description | Size | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Size | Price | Quantity & Availability | ||||
16217971
|
Fisherbrand™
|
Plain translucent bag, no print | 310 x 660 mm |
27.24 €
Pack of 100 |
In Stock
Log in to see stock. |
||||
16247971
|
Fisherbrand™
|
Printed with biohazard waste identification | 310 x 660 mm |
30.32 €
Pack of 100 |
In Stock
Log in to see stock. |
||||
16297961
|
Fisherbrand™
|
Printed with biohazard waste identification | 410 x 630 mm |
39.49 €
Pack of 100 |
Estimated Shipment: 10-07-2024
Log in to see stock. |
||||
16227971
|
Fisherbrand™
|
Plain translucent bag, no print | 410 x 630 mm |
36.94 €
Pack of 100 |
In Stock
Log in to see stock. |
||||
16207971
|
Fisherbrand™
|
Printed with biohazard waste identification | 620 x 780 mm |
69.96 €
Pack of 100 |
Estimated Shipment: 10-07-2024
Log in to see stock. |
||||
16237971
|
Fisherbrand™
|
Plain translucent bag, no print | 620 x 780 mm |
63.65 €
Pack of 100 |
In Stock
Log in to see stock. |
||||
Description
Designed to be strong, pliable, lean and puncture resistant. Fisherbrand™ Polypropylene Clear Autoclave Bags withstand high temperatures for autoclaving.
- For high temperature sterilisation (134°C) to a maximum of 140°C
- 40μm thickness
- High temperature polypropylene bags for the decontamination and inactivation of particularly resistant biological waste
- For safe disposal of biohazard waste
- Specifically designed for contaminated waste disposal in autoclaves or incinerators
- Printed with a biohazard warning
Specifications
Autoclavable | |
40 μm | |
134°C to 140°C |
Polypropylene | |
100/Pk. |