missing translation for 'onlineSavingsMsg'
Learn More

Ansell™ VersaTouch™ 92-465 Series Blue Nitrile Splash Gloves

Provides outstanding resistance to animal fats compared with PVC or natural rubber. Ansell VersaTouchTM 92-465 Series Blue Nitrile Splash Gloves are manufactured from an innovative nitrile compound which offers excellent chemical and mechanical properties, comfort and dexterity.

24.00 €

Specifications

Certifications/Compliance EN 420:2003 + I1:2009 and EN 374:2003
Material Nitrile
Thickness (English) 4.7 mil
Thickness (Metric) 0.12 mm
Series 92-465
View More Specs

Products 4
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
11707713
View Documents Promotion Details
Ansell™
92465070
6.5/7
24.00 €
Pack of 100
View alternative products
Save Up to 
»
Estimated Shipment: 02-07-2024
to see stock.
Add to basket
 
11717713
View Documents Promotion Details
Ansell™
92465090
8.5/9
24.00 €
Pack of 100
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
 
11727713
View Documents Promotion Details
Ansell™
92465100
9.5/10
24.00 €
Pack of 100
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
 
15670924
View Documents Promotion Details
Ansell™
92465080
7.5/8
24.00 €
100 pairs
View alternative products
Save Up to 
»
Estimated Shipment: 13-06-2024
to see stock.
Add to basket
 
Description

Description

  • Single use splash resistant gloves
  • Textured finish on fingertips. Improved grip
  • Contains no natural rubber
  • Suitable for contact with all fatty foods
  • Good resistance to animal fats compared with PVC or natural rubber
  • Powder-free
  • No risk of contaminating food with corn starch used to powder gloves

Applications:

  • All final stages of meat or fish processing
  • Controlled-environment rooms
  • Prepared meals
  • Dairy processing
  • Catering
Specifications

Specifications

EN 420:2003 + I1:2009 and EN 374:2003
4.7 mil
92-465
Nitrile
0.12 mm
Videos
SDS
Documents

Documents

Product Certifications