Learn More
Invitrogen™ Qubit™ Flex Verification Assay Kit
A fast, easy-to-use, reagent-based assay to test the performance of the Qubit™ Flex Fluorometer.
Brand: Invitrogen™ Q33254
58.96 EUR valid until 2024-06-28
Use promo code "21649" to get your promotional price.
Additional Details : Weight : 0.10000kg
Description
- Fast
- Easy-to-use
- Reagent-based assay
- Tests the performance of the Qubit™ Flex Fluorometer
- Consists of three components: A blank reagent solution, a green fluorescent reagent, and a far-red fluorescent reagent
- The green fluorescent reagent is designed to test blue excitation (470 nm LED; 430 to 495 nm excitation filter) and green fluorescence detection (510 to 580 nm emission filter)
- The far-red fluorescent reagent is designed to test red excitation (635 nm LED; 600 to 645 nm excitation filter) and far-red fluorescence detection (665 to 720 nm emission filter)
- Can only be used with the Qubit™ Flex Fluorometer
Specifications
Qubit Flex Fluorometer | |
Verification of Qubit Flex Fluorometer |
For Research Use Only. Not for use in diagnostic procedures.