missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eppendorf™ Research™ plus, Mechanical Single-Channel Pipettes Multi-Packs
Use these pipettes for stress-free work procedures and intuitive operation. Eppendorf™ Research™ plus Pipettes meet the highest needs in precision and accuracy – combined with ultimate ergonomics and increased flexibility.
790.00 € - 1660.00 €
Product Code | Brand | Includes | Volume (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | Volume (Metric) | Price | Quantity & Availability | ||||
15772069
|
Eppendorf™
3123000900 |
3 Pipettes: Research™ plus 1-Channel Variable, 3 x epT.I.P.S.™ Box 2.0 with 96 tips and ballpoint pen | 0.5 to 10, 10 to 100, 100 to 1000 μL |
790.00 €
Each |
In Stock
Log in to see stock. |
||||
15733459
|
Eppendorf™
3123000918 |
3 Pipettes: Research™ plus 1-Channel Variable, 3 x epT.I.P.S.™ Box 2.0 with 96 tips and ballpoint pen | 2 to 20, 20 to 200, 100 to 1000 μL |
790.00 €
1 set |
In Stock
Log in to see stock. |
||||
15743459
|
Eppendorf™
3123000926 |
3 Pipettes: Research™ plus 1-Channel Variable, 1 x epT.I.P.S.™ Box 2.0 with 96 tips, 2 x samples bags and ballpoint pen | 100 to 1000 μL, 0.5 to 5, 1 to 10 mL |
790.00 €
1 set |
In Stock
Log in to see stock. |
||||
17884601
|
Eppendorf™
3123000942 |
6 Pipettes: Research™ plus 1-Channel Variable, 1 x Pipette Carousel 2, 6 x epT.I.P.S.™ Box 2.0 with 96 tips and ballpoint pen | 0.1 to 2.5, 0.5 to 10, 2 to 20, 10 to 100, 20 to 200, 100 to 1000 μL |
1660.00 €
1 set |
In Stock
Log in to see stock. |
||||
Description
Durable and resistant to chemical, thermal and mechanical stress.
- Volume adjustment: only a few turns to reach desired volume
- Control button: very low operating force, color indicates pipet volume, positioned for perfect ergonomics
- Ejector: very low operating force, positioned for perfect ergonomics
- Volume display: four digits, magnifying shape
- Secondary adjustment display and opening: adjusts pipet to a liquid very different than water
- Eppendorf PerfectPiston™: ultra light piston made of Fortron™
- Quick connection clip: removes lower part easily (not in 10μl pipettes)
- Spring-loaded tip cone: improved ergonomics, tight fit to the tip
Spécification
Autoclavable | |
Ergonomic |