missing translation for 'onlineSavingsMsg'
Learn More

Cytiva (formerly Pall Lab) Sentino™ Funnel Base with Support Screen

Magnetic funnels replacement base designed for use with our Sentino pump.

Brand:  Cytiva (formerly Pall Lab) 4279

Additional Details : Weight : 0.26600kg

Product Code. 17194291

  • 222.00 € / Each
Estimated Shipment: 08-08-2024
to see stock.
Add to basket

Description

Description

  • Unique magnetic seal allows easy, one-handed vacuum filtration of liquids with compatible membranes
  • Not approved for use in medical, clinical, surgical or other patient applications
Specifications

Specifications

Polyphenylsulfone Stem and Support Screen
Non-sterile
Base for 150 and 300 mL
47 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers