Learn More
3M™ Specialty Particulate Respirator 9922
3M™ Specialty Particulate Respirator 9922 provides lightweight FFP2 protection against dusts, mists, ozone, and gases.
Brand: 3M™ 7000088787
49.85 EUR valid until 2024-12-31
Use promo code "22912" to get your promotional price.
Code : NEW
Additional Details : Weight : 1.82000kg
Description
- Provides FFP2 (NPF 12) protection against dusts, mists, ozone, and gases
- Activated carbon layer provides relief against nuisance-level odors
- 3M™ Cool Flow™ Valve helps reduce heat build-up
- Convex shape, nose clip and twin strapping helps provide a comfortable fit across a broad range of face shapes
- Lightweight, cup-shaped design is sturdy and collapse resistant
- Braided, color-coded headbands help identify the protection level offered: blue for FFP2
- Designed to work well with 3M™ Eyewear and Hearing Protection products
- Good fit over a wide range of face sizes provided by convex shape, nose clip and twin-strap design
Specifications
Gray | |
Specialty Particulate Respirator | |
<Hgv for ozone , Up to 10 x Hgv for particles |
FFP2 Protection Level | |
Yes |