Learn More
Merck Steritop™ E-GP Funnel-less Sterile Vacuum Filtration Systems
The newest addition to the Stericup™ Quick Release Family. Evolved with an eco-conscience.
219.00 €
Specifications
Color | Clear with blue adapter |
---|---|
Disposable | Single-use |
Material | Polystyrene |
Material (Membrane) | PES Express PLUS |
Pore Size | 0.22 μm |
Product Code | Brand | Outlet Fitting | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Outlet Fitting | Price | Quantity & Availability | |||||
16457545
|
Merck
SEGPT0038 |
38 mm threaded |
219.00 €
Pack of 12 |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||||
16467545
|
Merck
SEGPT0045 |
45 mm threaded |
219.00 €
Pack of 12 |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||||
Description
Stericup E and Steritop E sterile vacuum systems are the newest addition to the Stericup Quick Release Family. The Stericup E and Steritop E family of devices are designed as a more sustainable option for sterile filtration that significantly reduces plastic and packaging. These are funnel-less filter products for use in the sterile vacuum filtration of aqueous solutions such as media and tissue culture reagents/buffers.
The Stericup E and Steritop E devices are designed specifically for sterile filtration from compatible bottles (38 and 45 mm neck size) of media and tissue culture reagents/buffers.
SEGPT0038 is a 38 mm threaded collar, which uses a 0.22 μm pore size fast flow PES Express PLUS membrane that can directly connect to any volume, 45 mm receiver flask (glass or plastic) for sterilizing tissue culture media and buffers.
Specifications
Clear with blue adapter | |
Polystyrene | |
0.22 μm | |
40 cm² | |
Bottle Top Filter | |
73 mm |
Single-use | |
PES Express PLUS | |
Sterile | |
Sterile filtration | |
<2 mL (limited by vial volume) | |
73 mm |