missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ TaqPath™ qPCR Master Mix, CG
Designed and all lots are functionally tested to help ensure lot-to-lot reproducibility for Ct consistency and dynamic range across a wide variety of assays.
3050.00 € - 5940.00 €
Specifications
Content And Storage | Store in refrigerator (2° to 8°C). |
---|---|
Detection Method | Primer-probe |
For Use With (Equipment) | QuantStudio™ 12K Flex, QuantStudio™ 3, QuantStudio™ 5, QuantStudio™ 6, QuantStudio™ 7 |
Passive Reference Dye | ROX (Pre-mixed) |
For Use With (Application) | Real Time PCR (qPCR), Gene Expression, miRNA analysis |
Product Code | Brand | No. of Tests | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Tests | Price | Quantity & Availability | |||||
16268100
|
Applied Biosystems™
A16247 |
2500 Reactions |
3050.00 €
Pack of 5 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15820843
|
Applied Biosystems™
A15298 |
5000 Reactions |
5940.00 €
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Efficient and linear detection of up to 8 logs with gene expression or miRNA assays
- Enables reliable detection of low-copy templates with reproducible CT results
- Robust multiplexing performance with exogenous or endogenous targets
- Manufactured with stringent production and process controls to help ensure lot-to-lot consistency
- Labeled “For Laboratory Use”
Specifications
Store in refrigerator (2° to 8°C). | |
QuantStudio™ 12K Flex, QuantStudio™ 3, QuantStudio™ 5, QuantStudio™ 6, QuantStudio™ 7 | |
Real Time PCR (qPCR), Gene Expression, miRNA analysis |
Primer-probe | |
ROX (Pre-mixed) | |
DNA, cDNA |
For laboratory use.