missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TBE Buffer, Tris-Borate-EDTA, 10X Solution, Electrophoresis, Fisher BioReagents™
41.71 EUR valid until 2024-06-30
Use promo code "21742" to get your promotional price.
Use promo code "21742" to get your promotional price.
Click to view available options
Quantity:
1 L
4 L
20 L
Packaging:
Nalgene Poly Bottle
Poly CUBE
Description
TBE Buffer, Tris-Borate-EDTA, 10X Solution, Electrophoresis
- Save time and simplify your buffer preparation step by using Fisher BioReagents 10X TBE buffer solution - simply dilute as needed
- 10X solution contains 0.89M tris, 0.89M borate, and 0.02M EDTA
- To prepare a 1X solution, mix one volume of Fisher BioReagents 10X TBE Solution with 9 volumes of ultrapure water, such as BP2819
- Tested for the absence of DNase, RNase, and protease
- Functionally tested for use in electrophoresis applications
- TBE is a commonly used buffer for electrophoresis of nucleic acids in agarose and polyacrylamide gels
- Intended for laboratory research use only
Specifications
Color | Colorless |
pH | 8.2 to 8.4 |
Physical Form | Liquid |
Quantity | 1 L |
DNase | DNase free |
Filtered Through | Filtered through a 0.2-micron filter. |
Packaging | Nalgene Poly Bottle |
Protease | Protease free |
Grade | DNase-, RNase- and Protease-Free |
Chemical Name or Material | Tris-Borate-EDTA, 10X Solution |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'