Learn More
Epredia™ Shandon™ Sequenza™ Immunostaining Center Slide Rack
Slide Rack, for use with the Shandon™ Sequenza™ Immunostaining Center.
Brand:  Epredia™ 73310017
Code : 50
Additional Details : Weight : 0.36000kg
Product Code. 10098889
  • 616.00 € / Each
Estimated Shipment: 02-07-2024
to see stock.
Description
Description
  • Slide Rack holds 10 Shandon Coverplate™ assemblies, which ensure consistency and excellent staining quality
Specifications
Specifications
Slide Rack
Cassette with 10 staining positions
Shandon™ Sequenza™ Immunostaining Center