missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell TouchNTuff™ 92-600 Disposable Chemical Resistant Nitrile Gloves
20.19 EUR valid until 2024-06-30
Use promo code "21742" to get your promotional price.
20.19 EUR valid until 2024-12-31
Use promo code "21907" to get your promotional price.
Use promo code "21742" to get your promotional price.
20.19 EUR valid until 2024-12-31
Use promo code "21907" to get your promotional price.
Click to view available options
Size:
Small
Medium
Large
X-Large
Description
Soft nitrile disposable gloves provide high levels of comfort and splash resistance against hazardous chemicals.
- Made from 100% nitrile
- Resistant to nuisance punctures, nicks, cuts and abrasions
- Comfortable thin mil construction
- Rolled cuffs for easy donning and doffing
- Chemical resistant*
*Not recommended for Acetone, Benzaldehyde, Chlorobenzene, EDC, DMAC, DMF, Ethyl Acetate, Furfural, Methylene Bromide, MEK, Morpholine, NMP, Iodomethane, Nitric Acid -70%, (Concentrated), Nitric Acid, Red Fuming, Nitrobenzene, 1-Nitropropane, 2-Nitropropane, Phenol, 90%, THF, Pyridine, Sulfuric Acid, 95-98% (Concentrated) and THF
Specifications
Certifications/Compliance | ASTM D6319, FDA 21 CFR 177.2600, ASTM F1671 |
Certifications/Compliance (Europe) | Category III, EN 420:2003 + A1:2009 |
Chlorinated | No |
Color | Green |
Disposable | Single-use |
Finger Thickness (English) | 5.5 mil |
Finger Thickness (Metric) | 0.14 mm |
Hand | Ambidextrous |
Latex Free | Yes |
Material | Nitrile |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'