Learn More
Polysorbate 20, Fisher BioReagents™
Non-ionic detergent
Brand: Fisher Bioreagents BP337-100
17.60 EUR valid until 2024-06-30
Use promo code "21742" to get your promotional price.
Code : 32
Additional Details : CAS Number : 9005-64-5 Weight : 0.20868kg
Description
Non-ionic detergent
This nonionic detergent is used in the isolation of water-soluble membrane proteins.Specifications
Polysorbate 20 | |
9005-64-5 | |
100°C | |
(C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC18H34O6 | |
6 | |
Polyoxyethylene-20-sorbitan Monolaurate, Polyoxyethylenesorbitan monolaurate | |
CCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO | |
522.68 | |
Pass Test | |
0.25% max. | |
400 mPa/s at 25°C |
Amber | |
Viscous Liquid | |
1.100g/cm³ | |
MFCD00165986 | |
100 mL | |
HMFKFHLTUCJZJO-UHFFFAOYNA-N | |
2-[2-[3,4-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy]ethyl dodecanoate | |
96 to 108 | |
443314 | |
Glass Bottle | |
3% max. |