Learn More
HUBER Variostat™ Bath
Variostat™ Bath insulated stainless steel tanks are the ideal choice to incubate samples and keep them at a constant temperature.
Brand: HUBER 6052
Code : 04
Additional Details : Weight : 5.00000kg
Description
- Insulated, stainless steel tanks
- Drain port
- Bath cover sold separately
Specifications
5.5 L | |
210 mm | |
230 V 50 Hz | |
50 Hz |
30°C to 150°C | |
205 mm | |
230 V |