accessibility menu, dialog, popup

Test

CAS RN 100-11-8

ONOBr

CAS RN 100-11-8

IUPAC Name: 1-(bromomethyl)-4-nitrobenzene
Synonyms: nitrobenzyl bromide α-bromoparanitrotoluene
Molecular Weight (g/mol): 216.03
Molecular Formula: C7H6BrNO2
InChi Key: VOLRSQPSJGXRNJ-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(CBr)C=C1
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)