accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 100-76-5

CAS RN 100-76-5

IUPAC Name: 1-azabicyclo[2.2.2]octane
Synonyms: quinuclidine 1,4-ethylenepiperidine chinuclidin
Molecular Weight (g/mol): 111.19
Molecular Formula: C7H13N
InChi Key: SBYHFKPVCBCYGV-UHFFFAOYSA-N
SMILES: C1CN2CCC1CC2
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.